Skip to Content
MilliporeSigma
trans-1,2-Diaminocyclohexane-N,N,N′,N′-tetraacetic acid monohydrate puriss. p.a., ACS reagent, suitable for suitability for complexometry, ≥99.0% (KT)

trans-1,2-Diaminocyclohexane-N,N,N′,N′-tetraacetic acid monohydrate

trans-1,2-Diaminocyclohexane-N,N,N′,N′-tetraacetic acid monohydrate puriss. p.a., ACS reagent, suitable for suitability for complexometry, ≥99.0% (KT)
Synonym(s):
1,2-Cyclohexanedinitrilotetraacetic acid, Chel-CD, DCTA
Empirical Formula (Hill Notation):
C14H22N2O8 · H2O
CAS Number:
Molecular Weight:
364.35
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade