![Thulium foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%](/deepweb/assets/sigmaaldrich/product/structures/218/725/17717df8-2ddd-4cf1-a455-0bc192faa66a/640/17717df8-2ddd-4cf1-a455-0bc192faa66a.png)
Thulium
![Thulium foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%](/deepweb/assets/sigmaaldrich/product/structures/218/725/17717df8-2ddd-4cf1-a455-0bc192faa66a/640/17717df8-2ddd-4cf1-a455-0bc192faa66a.png)
Synonym(s):
Thulium
Empirical Formula (Hill Notation):
Tm
CAS Number:
Molecular Weight:
168.93
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Element
Showing 1-10 of 10
Compare
Product Number
Description
Pricing