Skip to Content
MilliporeSigma
Thimerosal European Pharmacopoeia (EP) Reference Standard

Thimerosal

Thimerosal European Pharmacopoeia (EP) Reference Standard
Synonym(s):
2-(Ethylmercuriomercapto)benzoic acid sodium salt, Ethylmercurithiosalicylic acid sodium salt, Mercury-([o-carboxyphenyl]thio)ethyl sodium salt, Sodium ethylmercurithiosalicylate, Thimerosal
Linear Formula:
2-(C2H5HgS)C6H4CO2Na
CAS Number:
Molecular Weight:
404.81

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Mode of Action
Package Size
API Family
Reference Material Type
Special Grade
Technique