Skip to Content
MilliporeSigma
Tetrakis(trimethylsilyl)silane ≥97.0%

Tetrakis(trimethylsilyl)silane

Tetrakis(trimethylsilyl)silane ≥97.0%
Synonym(s):
2,2-Bis(trimethylsilyl)-1,1,1,3,3,3-hexamethyl-trisilane
Linear Formula:
[(CH3)3Si]4Si
CAS Number:
Molecular Weight:
320.84

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing