Skip to Content
MilliporeSigma
Tetraethylene glycol monooctyl ether liquid, ≥98% (GC)

Tetraethylene glycol monooctyl ether

Tetraethylene glycol monooctyl ether liquid, ≥98% (GC)
Synonym(s):
C8E4, Octyl tetraethylene glycol ether, Octyltetraglycol
Linear Formula:
CH3(CH2)7(OCH2CH2)4OH
CAS Number:
Molecular Weight:
306.44

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing