Skip to Content
MilliporeSigma
Tetraethylene glycol dimethyl ether ≥99%

Tetraethylene glycol dimethyl ether

Tetraethylene glycol dimethyl ether ≥99%
Synonym(s):
2,5,8,11,14-Pentaoxapentadecane, Bis[2-(2-methoxyethoxy)ethyl] ether, Dimethoxytetraethylene glycol, Dimethyltetraglycol, Tetraglyme, Tetraethylene glycol dimethyl ether
Linear Formula:
CH3O(CH2CH2O)4CH3
CAS Number:
Molecular Weight:
222.28

Products

Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Functionality
Shape
Polymer Composition
Polymer Type
Reaction Type
Reagent Type