Skip to Content
MilliporeSigma
Tetradecyl-β-D-maltoside ≥99.0% (TLC)

Tetradecyl-β-D-maltoside

Tetradecyl-β-D-maltoside ≥99.0% (TLC)
Synonym(s):
n-Tetradecyl β-D-maltopyranoside
Empirical Formula (Hill Notation):
C26H50O11
CAS Number:
Molecular Weight:
538.67

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing