Skip to Content
MilliporeSigma
Tetrabutylammonium hydroxide 30-hydrate ≥98.0% (T)

Tetrabutylammonium hydroxide 30-hydrate

Tetrabutylammonium hydroxide 30-hydrate ≥98.0% (T)
Linear Formula:
(CH3CH2CH2CH2)4N(OH) · 30H2O
CAS Number:
Molecular Weight:
799.93

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Special Grade
Showing 1-2 of 2