Skip to Content
MilliporeSigma
tert-Butyl α-bromoisobutyrate ≥98.0%

tert-Butyl α-bromoisobutyrate

tert-Butyl α-bromoisobutyrate ≥98.0%
Synonym(s):
tert-Butyl 2-bromo-2-methylpropionate
Linear Formula:
(CH3)2CBrCOOC(CH3)3
CAS Number:
Molecular Weight:
223.11
EC Number:

Products

Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing