
Teriflunomide

Synonym(s):
(Z)-2-Cyano-3-hydroxy-N-[4-(trifluoromethyl)phenyl]-2-butenamide, A 1726, A77-1726, A771726, Flucyamide, HMR 1726, N-(4-Trifluoromethylphenyl)-2-cyano-3-hydroxycrotoamide, SU 20
Empirical Formula (Hill Notation):
C12H9F3N2O2
CAS Number:
Molecular Weight:
270.21
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Research Area
Quality Segment
API Family
Showing 1-2 of 2
Compare
Product Number
Description
Pricing