Skip to Content
MilliporeSigma
SUPELCOSIL™ LC-Si HPLC Column 3 μm particle size, L × I.D. 15 cm × 4.6 mm

SUPELCOSIL LC-Si (3 µm) HPLC Columns

SUPELCOSIL™ LC-Si HPLC Column 3 μm particle size, L × I.D. 15 cm × 4.6 mm

Products

Product Category
Brand
pH Value
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Application
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1