Skip to Content
MilliporeSigma
Sudan Orange G analytical standard

Sudan Orange G

Sudan Orange G analytical standard
Synonym(s):
2,4-Dihydroxyazobenzene, 4-(Phenylazo)resorcinol, SOG
Linear Formula:
C6H5N=NC6H3-1,3-(OH)2
CAS Number:
Molecular Weight:
214.22
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing