Skip to Content
MilliporeSigma
Stearidonic acid ≥99%

Stearidonic acid

Stearidonic acid ≥99%
Synonym(s):
Stearidonic acid, 6Z,9Z,12Z,15Z-Octadecatetraenoic acid, Moroctic acid, all-cis-6,9,12,15-Octadecatetraenoic acid
Empirical Formula (Hill Notation):
C18H28O2
CAS Number:
Molecular Weight:
276.41

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing