Skip to Content
MilliporeSigma
Stearic acid European Pharmacopoeia (EP) Reference Standard

Stearic acid

Stearic acid European Pharmacopoeia (EP) Reference Standard
Synonym(s):
1-Heptadecanecarboxylic acid, C18:0, Cetylacetic acid, NSC 25956, NSC 261168, Octadecanoic acid, Stearophanic acid, Stearic acid, Stearic Acid
Linear Formula:
CH3(CH2)16COOH
CAS Number:
Molecular Weight:
284.48

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique