Skip to Content
MilliporeSigma
Sphingomyelin from chicken egg yolk, ≥95%

Sphingomyelin

Sphingomyelin from chicken egg yolk, ≥95%
Synonym(s):
N-Acyl-4-sphingenyl-1-O-phosphorylcholine, N-Acyl-D-sphingosine-1-phosphocholine
CAS Number:
EC Number:

Products

Product Category
Brand
Biological Source
Physical Form
Purity
Tissue
Quality Segment
Functional Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing