Skip to Content
MilliporeSigma
(S)-(−)-Perillaldehyde technical grade, 92%

(S)-(−)-Perillaldehyde

(S)-(−)-Perillaldehyde technical grade, 92%
Synonym(s):
(−)-Perillaaldehyde, (S)-4-Isopropenyl-cyclohexene-1-carboxaldehyde, (S)-p-Mentha-1,8-dien-7-al
Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing