Skip to Content
MilliporeSigma
Sodium trifluoromethanesulfonate 98%

Sodium trifluoromethanesulfonate

Sodium trifluoromethanesulfonate 98%
Synonym(s):
Sodium triflate, Trifluoromethanesulfonic acid sodium salt
Linear Formula:
CF3SO3Na
CAS Number:
Molecular Weight:
172.06

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing