Skip to Content
MilliporeSigma
Sodium thiomethoxide solution 21% in H2O

Sodium thiomethoxide solution

Sodium thiomethoxide solution 21% in H2O
Synonym(s):
Sodium methanethiolate solution, Sodium thiomethoxide solution
Empirical Formula (Hill Notation):
CH3NaS
Molecular Weight:
70.09

Products

Product Category
Brand
Formula Weight
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing