Skip to Content
MilliporeSigma
Sodium salicylate European Pharmacopoeia (EP) Reference Standard

Sodium salicylate

Sodium salicylate European Pharmacopoeia (EP) Reference Standard
Synonym(s):
2-Hydroxybenzoic acid sodium salt, Salicylic acid sodium salt, Sodium 2-hydroxybenzoate, Sodium salicylate, Sodium Salicylate
Linear Formula:
HOC6H4COONa
CAS Number:
Molecular Weight:
160.10

Products

Product Category
Brand
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade