Skip to Content
MilliporeSigma
Sodium phosphate dibasic dodecahydrate SAJ first grade, ≥98.0

Sodium phosphate dibasic dodecahydrate

Sodium phosphate dibasic dodecahydrate SAJ first grade, ≥98.0
Synonym(s):
sec-Sodium phosphate, Disodium hydrogen phosphate dodecahydrate, Disodium phosphate, di-Sodium hydrogen phosphate dodecahydrate
Linear Formula:
Na2HPO4 · 12H2O
CAS Number:
Molecular Weight:
358.14
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Special Grade