Skip to Content
MilliporeSigma
Sodium perchlorate free-flowing, Redi-Dri™, ACS reagent, ≥98.0%

Sodium perchlorate

Sodium perchlorate free-flowing, Redi-Dri™, ACS reagent, ≥98.0%
Synonym(s):
Hyperchloric acid sodium salt, Sodium Perchlorate
Linear Formula:
NaClO4
CAS Number:
Molecular Weight:
122.44

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Application
Reagent Type
Special Grade