Skip to Content
MilliporeSigma
Sodium ethyl sulfate ≥98%

Sodium ethyl sulfate

Sodium ethyl sulfate ≥98%
Empirical Formula (Hill Notation):
C2H5NaO4S
CAS Number:
Molecular Weight:
148.11

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing