Skip to Content
MilliporeSigma
Sodium ethanethiolate technical grade, 80%

Sodium ethanethiolate

Sodium ethanethiolate technical grade, 80%
Synonym(s):
Ethanethiol sodium salt, Ethyl mercaptan sodium salt
Linear Formula:
CH3CH2SNa
CAS Number:
Molecular Weight:
84.12

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing