Skip to Content
MilliporeSigma
Sodium cyanoborohydride reagent grade, 95%

Sodium cyanoborohydride

Sodium cyanoborohydride reagent grade, 95%
Synonym(s):
Sodium cyanotrihydridoborate, Sodium cyanoborohydride
Linear Formula:
NaBH3CN
CAS Number:
Molecular Weight:
62.84

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Showing 1-3 of 3
Compare
Product Number
Description
Pricing