![Sodium cholesteryl sulfate](/deepweb/assets/sigmaaldrich/product/structures/785/842/87859e4c-fcb2-45fb-a349-73e35eea49e8/640/87859e4c-fcb2-45fb-a349-73e35eea49e8.png)
Sodium cholesteryl sulfate
![Sodium cholesteryl sulfate](/deepweb/assets/sigmaaldrich/product/structures/785/842/87859e4c-fcb2-45fb-a349-73e35eea49e8/640/87859e4c-fcb2-45fb-a349-73e35eea49e8.png)
Synonym(s):
5-Cholesten-3β-ol sulfate sodium salt, Cholesterol 3-sulfate sodium salt, Cholesteryl sodium sulfate, Cholesteryl sulfate sodium salt
Empirical Formula (Hill Notation):
C27H45NaO4S
CAS Number:
Molecular Weight:
488.70
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing