Skip to Content
MilliporeSigma
Sodium 1-heptanesulfonate Redi-Dri™, free-flowing

Sodium 1-heptanesulfonate

Sodium 1-heptanesulfonate Redi-Dri™, free-flowing
Synonym(s):
1-Heptanesulfonic acid sodium salt
Linear Formula:
C7H15O3SNa
CAS Number:
Molecular Weight:
202.25

Products

Product Category
Brand
Biological Source
Feature
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing