Skip to Content
MilliporeSigma
Sodium 1-decanesulfonate suitable for ion pair chromatography, LiChropur™, ≥99.0%

Sodium 1-decanesulfonate

Sodium 1-decanesulfonate suitable for ion pair chromatography, LiChropur™, ≥99.0%
Synonym(s):
1-Decanesulfonic acid sodium salt
Linear Formula:
CH3(CH2)8CH2SO3Na
CAS Number:
Molecular Weight:
244.33
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Technique