Skip to Content
MilliporeSigma
smartBoat™ disposable paper weighing boats size large, brown

smartBoat disposable paper weighing boats

smartBoat™ disposable paper weighing boats size large, brown
Synonym(s):
balance accessories, glassine weigh boat, glassine weighing paper, glassine weighing vessel, paper weigh boat, paper weighing vessel, plastic reduction weighing, white paper weigh boat

Products

Product Category
Color
Manufacturer
Height (mm)
Length (cm)
Length (mm)
Color
Package Size
Size