Skip to Content
MilliporeSigma
(S)-(−)-Limonene analytical standard

(S)-(−)-Limonene

(S)-(−)-Limonene analytical standard
Synonym(s):
(−)-p-Mentha-1,8-diene, (−)-Carvene, (S)-4-Isopropenyl-1-methyl cyclohexene
Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing