Skip to Content
MilliporeSigma
Silver lactate 97%

Silver lactate

Silver lactate 97%
Synonym(s):
Lactic acid silver salt
Linear Formula:
CH3CH(OH)COOAg
CAS Number:
Molecular Weight:
196.94

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing