(S)-(+)-Glycidyl methyl ether
Synonym(s):
(S)-(+)-2-(Methoxymethyl)oxirane, (S)-(+)-Methyl glycidyl ether
Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1