Skip to Content
MilliporeSigma
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine 97%

(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine

(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine 97%
Synonym(s):
(S)-(+)-1-AminoTetralin, (S)-(+)-1-Aminotetraline
Linear Formula:
(C10H11)NH2
CAS Number:
Molecular Weight:
147.22

Products

Product Category
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing