Skip to Content
MilliporeSigma
(−)-Scopolamine hydrobromide trihydrate ≥98% (HPLC), powder

(−)-Scopolamine hydrobromide trihydrate

(−)-Scopolamine hydrobromide trihydrate ≥98% (HPLC), powder
Synonym(s):
Hyoscine hydrobromide, Scopine tropate
Empirical Formula (Hill Notation):
C17H21NO4 · HBr · 3H2O
CAS Number:
Molecular Weight:
438.31

Products

Product Category
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing