Skip to Content
MilliporeSigma
(S)-(−)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid ≥99%

(S)-(−)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid

(S)-(−)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid ≥99%
Synonym(s):
(−)-MTPA, Mosher’s acid
Linear Formula:
C6H5C(OCH3)(CF3)CO2H
CAS Number:
Molecular Weight:
234.17
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing