Skip to Content
MilliporeSigma
Samarium(III) chloride hexahydrate ≥99.99% trace metals basis

Samarium(III) chloride hexahydrate

Samarium(III) chloride hexahydrate ≥99.99% trace metals basis
Synonym(s):
Samarium trichloride hydrate
Linear Formula:
SmCl3 · 6H2O
CAS Number:
Molecular Weight:
364.81

Products

Brand
Formula Weight
Physical Form
Purity
Reagent Type
Showing 1-2 of 2