Skip to Content
MilliporeSigma
(S)-(−)-1-Methyl-2-pyrrolidinemethanol 96%

(S)-(−)-1-Methyl-2-pyrrolidinemethanol

(S)-(−)-1-Methyl-2-pyrrolidinemethanol 96%
Synonym(s):
N-Methyl-L-prolinol, (S)-2-Hydroxymethyl-1-methylpyrrolidine
Empirical Formula (Hill Notation):
C6H13NO
CAS Number:
Molecular Weight:
115.17
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing