Skip to Content
MilliporeSigma
Rutin hydrate ≥94% (HPLC), powder

Rutin hydrate

Rutin hydrate ≥94% (HPLC), powder
Synonym(s):
Quercetin-3-rutinoside hydrate, Vitamin P hydrate
Empirical Formula (Hill Notation):
C27H30O16 · xH2O
CAS Number:
Molecular Weight:
610.52 (anhydrous basis)
EC Number:

Products

Product Category
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing