Skip to Content
MilliporeSigma
Ruthenium Red Technical grade

Ruthenium Red

Ruthenium Red Technical grade
Synonym(s):
Ammoniated ruthenium oxychloride
Linear Formula:
[(NH3)5RuORu(NH3)4ORu(NH3)5]Cl6
CAS Number:
Molecular Weight:
786.35

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing