Skip to Content
MilliporeSigma
(R)-Pantoic acid sodium salt ≥95% (GC)

(R)-Pantoic acid sodium salt

(R)-Pantoic acid sodium salt ≥95% (GC)
Synonym(s):
(2R)-2,4-Dihydroxy-3,3-dimethylbutanoic acid monosodium salt, (R)-2,4-Dihydroxy-3,3-dimethylbutyric acid sodium salt, Sodium (R)-pantoate, Sodium D-pantoate
Empirical Formula (Hill Notation):
C6H11NaO4
CAS Number:
Molecular Weight:
170.14

Products

Brand
Formula Weight
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing