Skip to Content
MilliporeSigma
Rosmarinic acid European Pharmacopoeia (EP) Reference Standard

Rosmarinic acid

Rosmarinic acid European Pharmacopoeia (EP) Reference Standard
Synonym(s):
(R)-O-(3,4-Dihydroxycinnamoyl)-3-(3,4- dihydroxyphenyl)lactic acid, 3,4-Dihydroxycinnamic acid (R)-1-carboxy-2-(3,4-dihydroxyphenyl)ethyl ester, Rosmarinic acid
Empirical Formula (Hill Notation):
C18H16O8
CAS Number:
Molecular Weight:
360.31

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique