Skip to Content
MilliporeSigma
Rosin phyproof® Reference Substance

Rosin

Rosin phyproof® Reference Substance
Synonym(s):
(2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(E)-3-phenylprop-2-enoxy]oxane-3,4,5-triol, Chinese rosin W, Rosin X, Trans-cinnamyl O-β-D-glucopyranoside
Empirical Formula (Hill Notation):
C15H20O6
CAS Number:
Molecular Weight:
296.32

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing