Skip to Content
MilliporeSigma
Ropinirole Related Compound B pharmaceutical secondary standard, certified reference material

Ropinirole Related Compound B

Ropinirole Related Compound B pharmaceutical secondary standard, certified reference material
Synonym(s):
4-[2-(Dipropylamino)ethyl]indoline-2,3-dione monohydrochloride
Empirical Formula (Hill Notation):
C16H22N2O2 · HCl
CAS Number:
Molecular Weight:
310.82

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade