Skip to Content
MilliporeSigma
(R)-(–)-3-Piperidinecarboxylic acid 97%

(R)-(–)-3-Piperidinecarboxylic acid

(R)-(–)-3-Piperidinecarboxylic acid 97%
Synonym(s):
(R)-(–)-Nipecotic acid
Empirical Formula (Hill Notation):
C6H11NO2
CAS Number:
Molecular Weight:
129.16

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing