Skip to Content
MilliporeSigma
Rilpivirine ≥98% (HPLC)

Rilpivirine

Rilpivirine ≥98% (HPLC)
Synonym(s):
4-[[4-[[4-[(1E)-2-Cyanoethenyl]-2,6-dimethylphenyl]amino]-2-pyrimidinyl]amino]benzonitrile, R 278474, TMC 278
Empirical Formula (Hill Notation):
C22H18N6
CAS Number:
Molecular Weight:
366.42

Products

Brand
Color
Feature
Formula Weight
Physical Form
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-2 of 2
Compare
Product Number
Description
Pricing