Skip to Content
MilliporeSigma
Rhodium(II) acetate dimer 99.9% trace metals basis

Rhodium(II) acetate dimer

Rhodium(II) acetate dimer 99.9% trace metals basis
Synonym(s):
Dirhodium tetraacetate, Tetrakis(acetato)dirhodium(II)
Linear Formula:
Rh2(OOCCH3)4
CAS Number:
Molecular Weight:
441.99
EC Number:

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing