Skip to Content
MilliporeSigma
Rhodamine 123 BioReagent, for fluorescence, ≥85% (HPLC)

Rhodamine 123

Rhodamine 123 BioReagent, for fluorescence, ≥85% (HPLC)
Synonym(s):
2-(6-Amino-3-imino-3H-xanthen-9-yl)benzoic acid methyl ester
Linear Formula:
C21H17ClN2O3
CAS Number:
Molecular Weight:
380.82
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing