Skip to Content
MilliporeSigma
Resorufin β-D-galactopyranoside ~95%

Resorufin β-D-galactopyranoside

Resorufin β-D-galactopyranoside ~95%
Synonym(s):
3-Phenoxazone 7-(β-D-galactopyranoside)
Empirical Formula (Hill Notation):
C18H17NO8
CAS Number:
Molecular Weight:
375.33

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing