Skip to Content
MilliporeSigma
vibrant-m

Recombinant Mouse IgG B2

vibrant-m

Products

Brand
Sustainability
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Quality Segment
Bottle material
Vial Bottle Material
Package Size
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing