Skip to Content
MilliporeSigma
(R)-(+)-1-Boc-3-aminopyrrolidine 97%

(R)-(+)-1-Boc-3-aminopyrrolidine

(R)-(+)-1-Boc-3-aminopyrrolidine 97%
Synonym(s):
(R)-(+)-N-Boc-3-aminopyrrolidine
Empirical Formula (Hill Notation):
C9H18N2O2
CAS Number:
Molecular Weight:
186.25

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing