Skip to Content
MilliporeSigma
(R)-(+)-1,1′-Bi(2-naphthol) 99%

(R)-(+)-1,1′-Bi(2-naphthol)

(R)-(+)-1,1′-Bi(2-naphthol) 99%
Synonym(s):
(+)-2,2′-Dihydroxy-1,1′-dinaphthyl, (R)-(+)-1,1′-Binaphthalene-2,2′-diol, (R)-BINOL
Linear Formula:
HOC10H6C10H6OH
CAS Number:
Molecular Weight:
286.32

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing